missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kainic acid monohydrate, 96%

Product Code. 15467999
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467999 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15467999 Supplier Thermo Scientific Chemicals Supplier No. J66590.MA
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58002-62-3 | C10H17NO5, C10H15NO4 | 231.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Description White to pale yellow
Content And Storage 2°C to 8°C
CAS 58002-62-3
Molecular Formula C10H15NO4
Formula Weight 213.23
Synonym 2-Carboxy-3-carboxymethyl-4-isopropenylpyrrolidine; (-)-(α)-Kainic Acid
Sensitivity Light Sensitive
Melting Point 251°C
Product Type Kainic Acid Monohydrate, 98%
Form Crystalline solid
Assay 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.