missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Kanamycin 1000μg Discs

Product Code. 13205269
Change view
Click to view available options
Quantity:
50 Discs/Cartridge (250 Discs/Pk.)
Unit Size:
Pack of 250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13205269 50 Discs/Cartridge (250 Discs/Pk.) Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13205269 Supplier Thermo Scientific™ Supplier No. DD0027B
  • ONLINE EXCLUSIVE PRICE
    123.00 €
    119.40 € / Pack of 250
    Save 3.60 €
    2% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Presumptively identify Gram-negative anaerobic bacilli using these diagnostic discs.

Presumptively identify Gram-negative anaerobic bacilli with Thermo Scientific™ Oxoid™ Kanamycin 1000μg Discs. Kanamycin is an aminoglycoside which, in high concentrations, can be used in combination with routine susceptibility discs to presumptively identify Gram-negative, anaerobic bacilli.

Use Kanamycin 1000μg Discs as a rapid and simple method for presumptive identification of anaerobic Gram-negative bacteria.

  • Rapid results
  • Simple and ready-to-use cartridge format.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C to +8°C
Description Kanamycin Discs
Concentration 1000 μg/disc
Detectable Analytes Anaerobic Gram-negative Bacteria
Format Disk, Cartridge
Type Kanamycin Disk
Packaging Type 5 Cart./Pk.
Quantity 50 Discs/Cartridge (250 Discs/Pk.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.