missing translation for 'onlineSavingsMsg'
Learn More

Kaolin, Pure, Light, Washed with Acid, Fisher Chemical™

Product Code. 10010220 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
3 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
3kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10010220 1 kg Plastic powder jar 1kg
10753991 3 kg Plastic keg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10010220 Supplier Fisher Chemical Supplier No. K/0050/60
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1332-58-7
Molecular Formula Al2H4O9Si2
Molecular Weight (g/mol) 258.16
InChI Key NLYAJNPCOHFWQQ-UHFFFAOYSA-N
Synonym kaolin, china clay, argilla, glomax, porcelain clay, bolus alba, white bole, hydrated aluminum silicate, kaolin clay slurry, alphagloss
PubChem CID 56841936
SMILES O.O.O=[Al]O[Si](=O)O[Si](=O)O[Al]=O

Specifications

Melting Point 1750°C
Color Beige
Physical Form Powder/Solid
Quantity 1 kg
MDL Number MFCD00062311
IUPAC Name oxo-oxoalumanyloxy-[oxo(oxoalumanyloxy)silyl]oxysilane;dihydrate
Formula Weight 258.2g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.