missing translation for 'onlineSavingsMsg'
Learn More

kappa-Carrageenan

Product Code. 15476588
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476588 25 g 25g
15466588 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476588 Supplier Thermo Scientific Alfa Aesar Supplier No. H60117.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 11114-20-8 | C24H36O25S2-2 | 788.647 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11114-20-8
Molecular Formula C24H36O25S2-2
Molecular Weight (g/mol) 788.647
MDL Number MFCD00151514
InChI Key ZNOZWUKQPJXOIG-XSBHQQIPSA-L
Synonym kappa-carrageenan, kappa-carrageenans, 1->4-3,6-anhydro-alpha-d-galactopyranosyl-1->3-4-o-sulfonato-beta-d-galactopyranan
PubChem CID 11966249
IUPAC Name [(2R,3S,4R,5R,6S)-6-[[(1R,3S,4R,5R,8S)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-[[(1R,3R,4R,5R,8S)-8-[(2S,3R,4R,5R,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-sulfonatooxyoxan-2-yl]oxy-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-3-yl]oxy]-5-hydroxy-2-(
SMILES C1C2C(C(O1)C(C(O2)O)O)OC3C(C(C(C(O3)CO)OS(=O)(=O)[O-])OC4C(C5C(C(O4)CO5)OC6C(C(C(C(O6)CO)OS(=O)(=O)[O-])O)O)O)O

Specifications

Color White
Quantity 25 g
Merck Index 14,1864
Formula Weight 401.32
Physical Form Solid
Chemical Name or Material kappa-Carrageenan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.