missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ KaryoStat™ HD Assay

Product Code. 17143019 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
24 assays
Unit Size:
24 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17143019 24 assays 24 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17143019 Supplier Applied Biosystems™ Supplier No. 905404
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Offers genotyping (sample ID) plus high resolution whole-genome coverage for accurate detection of chromosomal abnormalities

  • The KaryoStat HD Assay offers genotyping (sample ID) plus high resolution whole-genome coverage for accurate detection of chromosomal abnormalities. The assay has greater than 99% sensitivity and can reliably detect 25–50 kb copy number changes across the genome at high specificity with single-nucleotide polymorphism (SNP) allelic corroboration.
  • These high-density SNPs enable low-level mosaicism visualization and absence of heterozygosity (AOH), copy number change, and genomic contamination identification for research use. With more than 2.6 million copy number markers, the KaryoStat HD Assay covers all OMIM™ and RefSeq genes.
  • Our proprietary manufacturing technology produces arrays that are highly reproducible between each batch with no risk of probe dropout inherent in bead manufacturing techniques. The assay includes arrays, reagents, and easy-to-use data analysis software for a cost-effective and streamlined analysis of pluripotent stem cell lines.
  • Broadest coverage and highest performance for detecting chromosomal abnormalities
  • Karyotyping and genotyping (sample ID) with one assay
  • >99% sensitivity and can reliably detect 25–50 kb copy number changes
  • Simple analysis; no cytogenetics expertise necessary
  • Assay includes arrays, reagents
  • 2.5 days to results
TRUSTED_SUSTAINABILITY

Specifications

Type HD Assay
Number of Arrays 24 arrays
Format Genechip Probe Array
Species Human
Content And Storage Store at 2–8°C:
• Arrays
• Module 2: Pre-Lab TE Buffer and Water
• Module 4: Main Lab Stain, Holding Buffer, Beads, and Water

Store at -5 to -30°C:
• Module 3: Main Lab Fragmentation, Labeling, and Hybridization

Store at room temperature:
• Module 5: Main Lab Elution Buffer
• Purification Wash Buffer
• Wash Buffer A
• Wash Buffer B

For Use With (Equipment) GeneChip Scanner 3000
Includes Arrays, Reagents, Data Analysis Software
Product Line KaryoStat™ HD
Quantity 24 assays
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.