missing translation for 'onlineSavingsMsg'
Learn More

Nordvlies Katrin Basic System Toilet

Product Code. 15700937
Change view
Click to view available options
Color:
White
Unit Size:
36 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15700937 White 36 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15700937 Supplier Nordvlies Supplier No. 15615
  • ONLINE EXCLUSIVE PRICE
    105.00 €
    52.92 € / 36 rolls
    Save 52.08 €
    49% Off
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KATRIN Basic System Toilet - Economical and effective toilet paper for every day use.

  • Economical and effective toilet paper for every day use
  • 100% recycled fiber
  • 1-Ply, natural white
  • For use with Katrin System Toilet Dispenser
  • Suitable for high traffic areas
  • Controlled dispensing, and self presenting sheets reduce consumption and improve hygiene
  • All Katrin toilet papers are made from high quality fibers and have been proven, via dissolving tests, to break down within 90 seconds
  • Certified under the Nordic Swan Eco Label

Specifications

Certifications/Compliance Nordic Swan Eco Label
Color White
For Use With (Equipment) Katrin System Toilet Dispenser
Ply 1-ply
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.