missing translation for 'onlineSavingsMsg'
Learn More

Nordvlies Katrin Plus System Toilet 3

Product Code. 15771097
Change view
Click to view available options
Color:
White
Unit Size:
36 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15771097 White 36 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15771097 Supplier Nordvlies Supplier No. 968
  • ONLINE EXCLUSIVE PRICE
    127.00 €
    99.15 € / 36 rolls
    Save 27.85 €
    21% Off
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plus System Toilet 3 is high quality, soft effective toilet paper.

  • High quality, soft effective toilet paper
  • 3-Ply, white
  • For use with Katrin System Toilet Dispenser
  • Has that little extra in terms of gentleness, absorbency and strength
  • Suitable for high traffic areas in more exclusive public washrooms
  • Controlled dispensing, and self presenting sheets reduce consumption and improve hygiene
  • All Katrin toilet papers are made from high quality fibers and have been proven, via dissolving tests, to break down within 90 seconds
  • Certified under the Nordic Swan Eco Label and EU Flower

Specifications

Product Type Toilet Paper
Certifications/Compliance Nordic Swan Eco Label, EU Flower
Color White
For Use With (Equipment) Katrin System Toilet Dispenser
Ply 3-ply
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.