missing translation for 'onlineSavingsMsg'
Learn More

Nordvlies Katrin Plus Toilet 250

Product Code. 15760907
Change view
Click to view available options
Color:
White
Unit Size:
48 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15760907 White 48 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15760907 Supplier Nordvlies Supplier No. 10487
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Plus Toilet 250 Strong, high quality conventional toilet rolls

  • Strong, high quality conventional toilet rolls
  • 3-Ply, white, 250 sheets
  • Soft, absorbent paper for maximum comfort
  • Especially suitable for more exclusive public environments
  • All Katrin toilet papers are made from high quality fibers and have been proven, via dissolving tests, to break down within 90 seconds
  • Certified under the Nordic Swan Eco Label
  • Certified under the EU Flower Eco Label

Specifications

Product Type Toilet Paper
Certifications/Compliance Nordic Swan Eco Label, EU Flower Eco Label
Color White
For Use With (Equipment) Katrin System Toilet Dispenser
Packaging Quantity 250 Sheets
Ply 3-ply
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.