missing translation for 'onlineSavingsMsg'
Learn More

KD Scientific Legato™ Series Adapter for Hamilton™ Syringes

Product Code. 11935596
Change view
Click to view available options
For Use With (Equipment):
Gastight Syringe (25 and 50 mL)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11935596 Gastight Syringe (25 and 50 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11935596 Supplier KD Scientific Supplier No. 788314
  • ONLINE EXCLUSIVE PRICE
    40.55 €
    35.22 € / Pack of 1
    Save 5.33 €
    13% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For 25, 50 and 100 mL Hamilton™ Gastight™ Syringe

  • Selection of accessories and replacement parts for your Legato™ Series Pumps
  • Required for withdraw function

Specifications

Latex Free Yes
Product Type Adapter
Disposable Reusable
For Use With (Equipment) Gastight Syringe (25 and 50 mL)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.