missing translation for 'onlineSavingsMsg'
Learn More

KD Scientific Stainless-Steel Syringes

Product Code. 12867362
Change view
Click to view available options
Volume (Metric):
2.5 mL
8 mL
20 mL
50 mL
100 mL
200 mL
Product Type:
Luer-Lok Syringe
Swagelok 1/16 in. Syringe
Swagelok 1/4 in. Syringe
Swagelok 1/8 in. Syringe
Unit Size:
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Product Type unitSize
12867362 2.5 mL Swagelok 1/16 in. Syringe Each
12887362 8 mL Swagelok 1/16 in. Syringe Each
12897362 20 mL Swagelok 1/16 in. Syringe Each
15164174 20 mL Swagelok 1/8 in. Syringe Each
15194174 20 mL Swagelok 1/4 in. Syringe Each
15144174 8 mL Swagelok 1/8 in. Syringe Each
11951017 20 mL Luer-Lok Syringe Each
11986706 50 mL Swagelok 1/16 in. Syringe Each
13406038 50 mL Swagelok 1/8 in. Syringe Each
15174174 100 mL Swagelok 1/8 in. Syringe Each
15104184 50 mL Swagelok 1/4 in. Syringe Each
11945596 100 mL Swagelok 1/16 in. Syringe Each
11971017 100 mL Luer-Lok Syringe Each
15114184 100 mL Swagelok 1/4 in. Syringe Each
11961017 50 mL Luer-Lok Syringe Each
17129280 200 mL Swagelok 1/8 in. Syringe Each
11905547 200 mL Swagelok 1/4 in. Syringe Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 12867362 Supplier KD Scientific Supplier No. 780801
Add to basket
Add to basket
This item is not returnable. View return policy

Durable, electron-beam construction withstands high force, high pressure applications

Eliminates hazards of glass syringe breakage.

  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals

Specifications

Autoclavable Autoclavable
Material Stainless Steel
Diameter (English) Inner 19/100 in.
Diameter (English) Outer 1/2 in.
Diameter (Metric) Inner 4.8 mm
Diameter (Metric) Outer 12.7 mm
Product Type Swagelok 1/16 in. Syringe
Max. Pressure 7000 psi
Volume (Metric) 2.5 mL
For Use With (Equipment) Syringe Pump
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.