missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KD Scientific Stainless-Steel Syringes
Click to view available options
Volume (Metric):
2.5 mL
8 mL
20 mL
50 mL
100 mL
200 mL
Product Type:
Luer-Lok Syringe
Swagelok 1/16 in. Syringe
Swagelok 1/4 in. Syringe
Swagelok 1/8 in. Syringe
Unit Size:
Each
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Diameter (English) Inner | 1-37/100 in. |
| Diameter (English) Outer | 1-1/5 in. |
| Diameter (Metric) Inner | 34.7 mm |
| Diameter (Metric) Outer | 38.1 mm |
| Product Type | Swagelok 1/8 in. Syringe |
| Max. Pressure | 1500 psi |
| Volume (Metric) | 100 mL |
| For Use With (Equipment) | Syringe Pump |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction