missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ KECK™ Clip, for conical joints, from POM

Product Code. 11699808 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
NS 12
NS 14
NS 19
NS 24
NS 29
NS 34
NS 40
NS 45
Color:
Amber
Blue
Green
Orange
Red
Violet
Yellow
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Color Unit Size
11699808 NS 29 Red Pack of 10
10643353 NS 14 Yellow Pack of 10
10404672 NS 19 Blue Pack of 10
10011242 NS 24 Green Pack of 10
10571803 NS 19 Green Pack of 10
10264632 NS 12 Violet Pack of 10
10749474 NS 34 Orange Pack of 10
10254682 NS 45 Amber Pack of 10
10592183 NS 40 Yellow Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11699808 Supplier DWK Life Sciences™ Supplier No. 1091142
  • ONLINE EXCLUSIVE PRICE
    54.89 €
    50.49 € / Pack of 10
    Save 4.40 €
    8% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The KECK™ clamp is used to secure a standard ground joint connection.

KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).

  • For conical joints
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to set up
  • Increased safety in the laboratory as the glass surface is not scratched
  • Marking of the individual sizes for better differentiation

Specifications

Quantity 10
Material POM
For Use With (Equipment) Secure Glass-to-Glass Joints
For Use With (Application) Secure glass-to-glass joints
Size NS 29
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.