Learn More
Thermo Scientific Chemicals Kemptide
For cellular and molecular biology applications
Brand: Thermo Scientific Chemicals J60591.#0
Code : NEW
Additional Details : CAS Number : 65189-71-1 Weight : 0.00005kg
Description
Kemptide, phosphate acceptor, is shown to be a basic serine-containing heptapeptide corresponding to a sequence from pig liver pyruvate kinase. This synthetic peptide is a substrate of PKA (cyclic AMP-dependent protein kinase) and is phosphorylated and activated after microinjection into Xenopus oocytes. Exposure of Kemptide to insulin has demonstrated an increase in activation of this agent, which can be inhibited by the catalytic subunit of phosphatase 2A.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
-30°C to -10°C | |
Synthetic peptide substrate for protein kinase A | |
Kemptide | |
RUO - research use only |
Phosphate acceptor peptide | |
Powder | |
771.91 | |
5 mg | |
Leu-Arg-Arg-Ala-Ser-Leu-Gly |
RUO – Research Use Only