missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Kenpaullone
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
- Kenpaullone is an ATP-competitive inhibitor of glycogen synthase kinase 3β (GSK-3β; Bain et al.; Leclerc et al.; Zaharevitz et al.) It inhibits GSK-3β, Cdk1/cyclin B, Cdk2/cyclin A, Cdk5/p25, and lymphocyte kinase with IC50 values of 0.23, 0.4, 0.68, 0.85, and 0.47 μM, respectively (Bain et al.; Zaharevitz et al.)
- Replaces Klf4 in the generation of induced pluripotent stem (iPS) cells from mouse embryonic fibroblasts (MEFs) transduced with Oct4, Sox2, and c-Myc (Lyssiotis et al.)
- Enhances neuronal differentiation in rat and human neural precursor cell cultures (Castelo-Branco et al.; Lange et al.)
- Promotes survival of motor neurons derived from mouse embryonic stem (ES) cells and from Amyotrophic Lateral Sclerosis (ALS) patient iPS cells (Yang et al.)
- Inhibits KLF4 expression and self-renewal in breast cancer stem cells in vitro (Yu et al.)
Specifications
Specifications
| CAS | 142273-20-9 |
| Synonym | 9-Bromopaullone; NSC 664704 |
| Quantity | 10 mg |
| Molecular Formula | C16H11BrN2O |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction