missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, purum p.a, Honeywell™

Product Code. 15626000 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15626000

Brand: Honeywell Research Chemicals 607102.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 8008-20-6
MDL Number MFCD00135561

Specifications

Color Colorless
Density 0.8g/mL at 25°C
Boiling Point 175°C to 325°C
Packaging Glass bottle
Quantity 2.5 L
UN Number UN1223
Vapor Density 4.5 (vs air)
Physical Form Liquid
Chemical Name or Material Kerosene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.