missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, purum p.a, Honeywell™

Product Code. 15636000 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
5 L
Product Code. Quantity Packaging unitSize
15636000 5 L Glass bottle 5 L
15616000 1 L Glass bottle 1 L
15626000 2.5 L Glass bottle 2.5 L
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15636000

Brand: Honeywell Research Chemicals 607105L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Chemical Identifiers

CAS 8008-20-6
MDL Number MFCD00135561

Specifications

Color Colorless
Density 0.8g/mL at 25°C
Boiling Point 175°C to 3257deg;C
Packaging Glass bottle
Quantity 5 L
UN Number UN1223
Vapor Density 4.5 (vs air)
Physical Form Liquid
Chemical Name or Material Kerosene
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Kerosene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Kerosene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.