missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Ketoconazole
Description
Inhibitor of cytochrome P450c17. Enhances the formation of myelin basic protein-positive oligodendrocytes from oligodendrocyte progenitor cells in vitro and promotes remyelination in vivo. Also an antifungal agent. Brain penetrant.
Specifications
Specifications
| CAS | 65277-42-1 |
| Quantity | 100 mg |
| Product Line | Tocris |
| Molecular Formula | C26H28Cl2N4O4 |
| Formula Weight | 531.44 |
| Purity | >99% |
| Molecular Weight (g/mol) | 534.452 |
| Chemical Name or Material | Ketoconazole |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?