missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ketoconazole, 98%

Product Code. 15525156 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15525156 25 g 25g
15321788 1 g 1g
15515156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15525156 Supplier Thermo Scientific Chemicals Supplier No. 455470250
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ketoconazole, CAS # 65277-42-1, is a synthetic, broad-spectrum antifungal agent with the ability to inhibit a variety of fungal infections. | CAS: 65277-42-1 | C26H28Cl2N4O4 | 531.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ketoconazole is an imidazole antifungal agent
  • It functions by inhibiting synthesis of ergosterol, which is the fungal equivalent of cholesterol

Applications

  • Ketoconazole shows fungicide activity against a variety of yeast infections
  • It is an inhibitor of CYP3A4, which is necessary for the conversion of lanosterol to ergosterol
  • It increases fungal cellular permeability
  • It also inhibits endogenous respiration by disrupting membrane phospholipids
  • Ketoconazole can also inhibit the synthesis of thromboxane and sterols such as aldosterone, cortisol, and testosterone
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 531.44
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym ketoconazole
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Melting Point 148.0°C to 150.0°C
Loss on Drying 0.5% max. (80°C, 4 h) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in methanol,less soluble in ethanol,slightly soluble in dichloromethane or chloroform
Formula Weight 531.44
Percent Purity 98%
Chemical Name or Material Ketoconazole
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ketoconazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360F-May damage fertility.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.