missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, Fisher BioReagents

Product Code. 11448353
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
1 g
Packaging:
Amber Glass
Unit Size:
100mg
1g
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11448353 1 g Amber Glass 1g
10430015 50 mg Amber Glass 50mg
10754344 100 mg Amber Glass 100mg
10032624 500 mg Amber Glass 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11448353 Supplier Fisher BioReagents Supplier No. BP27341

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Ketoconazole is a cytochrome P450 inhibitor and demonstrates anti-psoriatic activity. An antifungal agent.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 534.452
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym ketoconazole
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Spezifikation

Melting Point 148°C
Color White
Ignition Residue < 0.1%
Loss on Drying < 0.5%
Melting Range Inclusive between 148°C
Assay Percent Range 98%
Packaging Amber Glass
Quantity 1 g
Merck Index 15, 5349
Formula Weight 531.44
Physical Form Solid
Chemical Name or Material Ketoconazole
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Ketoconazole
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Gefahrenhinweis
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360F-May damage fertility.
  • H373-May cause damage to organs.
  • H301-Toxic if swallowed.
Sicherheitshinweis
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.