missing translation for 'onlineSavingsMsg'
Learn More

α-Ketoglutaric Acid, MP Biomedicals

Product Code. 11436435 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436435 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11436435 Supplier MP Biomedicals Supplier No. 0520176705
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 328-50-7
Molecular Formula C5H6O5
MDL Number MFCD00004165
Quantity 5 g
Synonym 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
InChI Key KPGXRSRHYNQIFN-UHFFFAOYSA-N
SMILES C(CC(=O)O)C(=O)C(=O)O
IUPAC Name 2-oxopentanedioic acid
Molecular Weight (g/mol) 146.098
PubChem CID 51
ChEBI CHEBI:30915
Chemical Name or Material Alpha-Ketoglutaric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.