missing translation for 'onlineSavingsMsg'
Learn More

alpha-Ketoglutaric acid sodium salt, 98%

Product Code. 11965111 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11965111 25 g 25g
10399569 5 g 5g
11975111 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11965111 Supplier Thermo Scientific Acros Supplier No. 439350250
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 22202-68-2 | C5H5NaO5 | 168.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22202-68-2
Molecular Formula C5H5NaO5
Molecular Weight (g/mol) 168.08
InChI Key MOTOGHHLNTXPTI-UHFFFAOYSA-M
Synonym sodium hydrogen 2-oxoglutarate, pentanedioic acid, 2-oxo-, sodium salt, pentanedioic acid, 2-oxo-, sodium salt 1:?, 2-ketoglutaric acid monosodium salt, 2-oxoglutaric acid, sodium salt, glutaric acid, 2-oxo-, sodium salt, alpha-ketoglutaric acid, sodium salt, 2-oxoglutaric acid 1-sodium salt, sodium 4-carboxy-4-oxobutanoate
PubChem CID 23672314
IUPAC Name sodium;5-hydroxy-4,5-dioxopentanoate
SMILES C(CC(=O)[O-])C(=O)C(=O)O.[Na+]

Specifications

Color White
CAS Min % 97.5
CAS Max % 100.0
pH 3.6 to 4.0 (0.5% aq. soln.)
Physical Form Powder
Quantity 25 g
Assay Percent Range 98%
Linear Formula HO2CCH2CH2C(=O)CO2Na
Formula Weight 168.08
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 1.0% max.
Packaging Glass Bottle
Chemical Name or Material α-Ketoglutaric acid sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • alpha-Ketoglutaric acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.