missing translation for 'onlineSavingsMsg'
Learn More

Ketorolac tromethamine salt, MedChemExpress

Product Code. 16442059
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442059 1 g 1g
16420788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442059 Supplier Medchem Express Supplier No. HYB0138
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ketorolac tromethamine salt (RS37619 tromethamine salt) is a non-steroidal anti-inflammatory agent, acting as a nonselective COX inhibitor, with IC50s of 20 nM for COX-1 and 120 nM for COX-2.

Ketorolac tromethamine salt (RS37619 tromethamine salt) is a non-steroidal anti-inflammatory agent, acting as a nonselective COX inhibitor, with IC50s of 20 nM for COX-1 and 120 nM for COX-2.

Specifications

Chemical Name or Material Ketorolac tromethamine salt
CAS 74103-07-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H24N2O6
Quantity 1 g
Synonym Ketorolac Tromethamine Ketorolac tris salt RS37619 tromethamine salt
Solubility Information H2O : 100 mg/mL (265.67 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (79.70 mM)
SMILES O=C(C1C2=CC=C(C(C3=CC=CC=C3)=O)N2CC1)O.OCC(CO)(N)CO
Molecular Weight (g/mol) 376.4
Formula Weight 376.4
Percent Purity 99.52%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.