missing translation for 'onlineSavingsMsg'
Learn More

Ketotifen Fumarate, TRC

Product Code. 17980955
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17980955 500 mg 500mg
17982315 250 mg 250mg
17962745 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17980955 Supplier Toronto Research Chemicals Supplier No. TRCK315100500MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34580-14-8
Molecular Formula C19 H19 N O S . C4 H4 O4
Molecular Weight (g/mol) 425.5
Synonym Ketotifen Hydrogen Fumarate, 10H-Benzo[4,5]cyclohepta[1,2-b]thiophen-10-one, 4,9-dihydro-4-(1-methyl-4-piperidinylidene)-, (2E)-2-butenedioate (1:1), 10H-Benzo[4,5]cyclohepta[1,2-b]thiophen-10-one, 4,9-dihydro-4-(1-methyl-4-piperidinylidene)-, (E)-2-butenedioate (1:1), Allerkif, HC 20511, HC 20511 fumarate, Ketotifen fumarate, Ketotifen hydrogen fumarate, Totifen, Zaditen, Zaditor, Zasten
IUPAC Name (E)-but-2-enedioic acid;10-(1-methylpiperidin-4-ylidene)-5H-benzo[1,2]cyclohepta[2,4-b]thiophen-4-one
SMILES CN1CCC(=C2c3ccccc3CC(=O)c4sccc24)CC1.OC(=O)\C=C\C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H19NOS.C4H4O4/c1-20-9-6-13(7-10-20)18-15-5-3-2-4-14(15)12-17(21)19-16(18)8-11-22-19;5-3(6)1-2-4(7)8/h2-5,8,11H,6-7,9-10,12H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+
Formula Weight 425.1297
Percent Purity >95%
Chemical Name or Material Ketotifen Hydrogen Fumarate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.