missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional Key Set A 838

Product Code. 30130363 Shop All Miele Products
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30130363 Stainless Steel 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30130363 Supplier Miele Professional Supplier No. 11054290
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A 838 Key set for adjusting basket heights and installing/removing injector nozzles.

  • Set consisting of Torx wrench and open spanner
  • Suitable for Miele upper baskets and mobile units
TRUSTED_SUSTAINABILITY

Specifications

Product Type L-shaped Hex Key Set
Material Stainless Steel
Color Silver
For Use With (Application) Setting basket heights and assembling/dismantling injector nozzles
Includes Torx Wrench and Open Wrench
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.