Learn More
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format
Product Code. 17140069 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
24 Combi tip comb
24 DW plate, barcoded, sterile
24 DW tip comb and plate
24 DW tip comb, plate, barcode
24 DW tip comb, plate, sterile
24 Deep Well plate, barcoded
24 Deep-well Plate
24 Deep-well Plate, Sterile
6-tip comb and 24 DW plate
For Use With (Equipment):
KF Duo Prime, Flex, Presto
KingFisher Apex
KingFisher Duo Prime
KingFisher Flex and Presto
Unit Size:
Pack of 48
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) unitSize
17140069 24 Deep Well plate, barcoded KingFisher Apex Pack of 50
16552452 24 Combi tip comb KingFisher Apex Pack of 50
12653116 24 Deep-well Plate KF Duo Prime, Flex, Presto Pack of 50
12663116 24 Deep-well Plate, Sterile KF Duo Prime, Flex, Presto Pack of 50
12624075 24 DW tip comb and plate KingFisher Flex and Presto Pack of 50
12643116 6-tip comb and 24 DW plate KingFisher Duo Prime Pack of 48
17190069 24 DW tip comb, plate, barcode KingFisher Apex Pack of 50
17150069 24 DW plate, barcoded, sterile KingFisher Apex Pack of 50
17100079 24 DW tip comb, plate, sterile KingFisher Apex Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 17140069 Supplier Thermo Scientific™ Supplier No. 95040470B
Add to basket
Add to basket
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Product Type 24 Deep Well plate, barcoded
For Use With (Equipment) KingFisher Apex
Quantity 50/Cs.