missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ KingFisher Apex Accessories

Product Code. 18466738 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
24 Combi Head
24 DW Heating Block
96 DW Head
96 DW Heating Block
96 KF Heating Block
96 PCR Head
Combi Head
Heating Block 24
Heating Block 96
Matrix Heating Block
PCR Plate Heating Block
Plate Adapter
Storage Tube Adapter
UV Lamp (Replacement)
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
18466738 Matrix Heating Block Each
16552442 96 PCR Head Each
16562442 Combi Head Each
16572442 96 DW Head Each
16582442 24 Combi Head Each
16592432 PCR Plate Heating Block Each
16502442 96 KF Heating Block Each
16512442 96 DW Heating Block Each
16522442 24 DW Heating Block Each
16532442 Heating Block 96 Each
16542442 Heating Block 24 Each
16572452 Storage Tube Adapter Each
16582452 Plate Adapter Each
16592452 UV Lamp (Replacement) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 18466738 Supplier Thermo Scientific™ Supplier No. 24075970
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source.

KingFisher Apex is a flexible system for different types of sample processing needs which offers highly versatile, automated magnetic-particle processing for DNA/RNA, protein and cell purification from virtually any source. Using revolutionary magnetic particle separation technology, this system provides excellent reproducibility and quality

  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval
TRUSTED_SUSTAINABILITY

Specifications

Product Type Matrix Heating Block
Disposable Reusable
For Use With (Application) DNA/RNA, protein and cell purification
For Use With (Equipment) KingFisher Apex
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.