Learn More
KGW Isotherm™ KGW Isotherm
Product Code. 15311271
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material Unit Size
15311271 Stainless Steel Each
1 options
Product Code. 15311271 Supplier KGW Isotherm™ Supplier No. 17111
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Capacity (Metric) 200 mL (Condensate), 1.2 L (Coolant)
Material Stainless Steel
Quantity 1 Pack
Type Cold Trap