missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Khellin, 95%

Product Code. 16534214 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16534214 25 g 25g
16524214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16534214 Supplier Thermo Scientific Chemicals Supplier No. 461800250
 Restricted Item
Estimated Shipment: 01-12-2026
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 82-02-0 | C14H12O5 | 260.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 82-02-0
Molecular Formula C14H12O5
Molecular Weight (g/mol) 260.25
InChI Key HSMPDPBYAYSOBC-UHFFFAOYSA-N
IUPAC Name 4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one
SMILES COC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O

Especificaciones

Melting Point 150.0°C to 154.0°C
Assay Percent Range 95%
Quantity 25 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 260.24
Chemical Name or Material Khellin

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.