missing translation for 'onlineSavingsMsg'
Learn More

Khellin, TRC

Product Code. 30179033
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179033 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179033 Supplier Toronto Research Chemicals Supplier No. TRCK4320001G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 82-02-0
Molecular Formula C14 H12 O5
Quantity 1 g
Synonym 5H-Furo[3,2-g][1]benzopyran-5-one, 4,9-dimethoxy-7-methyl-, 4,9-Dimethoxy-7-methyl-5-oxo-1,8-dioxabenz[f]indene, 4,9-Dimethoxy-7-methyl-5-oxofuro[3,2-g]-1,2-chromene, 4,9-Dimethoxy-7-methyl-5-oxofuro[3,2-g][1]benzopyran, 4,9-Dimethoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one, 5,8-Dimethoxy-2-methyl-4',5'-furano-6,7-chromone, 5,8-Dimethoxy-2-methyl-4',5'-furo-6,7-chromone, 5,8-Dimethoxy-2-methyl-6,7-furanochromone, Amicardien, Amicardine, Amikellin, Ammipuran, Ammispasmin, Ammivin, Ammivisnagen, Benecardin, Benekardin, Cardio-khellin, Corafurone, Coronin, Eskel, Gynokhellan, Gynokhellin, Hkelfren, Kalangin, Kelamin, Kelicor, Kelicorin, Kellosal, Keloid, Khelfren, Khelisem, Khellin, Khelline I, Khelloyd, Lynamine, Medekellin, Methafrone, NSC 25509, NSC 37744, NSC 8519, Norkel, Simeskellina, Vasokellina, Visammin, Viscardan, Visnagalin, Visnagen
InChI Formula InChI=1S/C14H12O5/c1-7-6-9(15)10-11(16-2)8-4-5-18-12(8)14(17-3)13(10)19-7/h4-6H,1-3H3
SMILES COc1c2OC(=CC(=O)c2c(OC)c3ccoc13)C
IUPAC Name 4,9-dimethoxy-7-methylfuro[3,2-g]chromen-5-one
Molecular Weight (g/mol) 260.24
Formula Weight 260.0685
Chemical Name or Material Khellin (Visammin)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.