missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KHS 101 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
KHS 101 hydrochloride is a selective inducer of neuronal differentiation; induces neuronal differentiation in cultured hippocampal neural progenitor cells (NPCs) by interacting with TACC3 (EC50 ∽1 μM). Suppresses astrocyte formation. Also induces acceleration of neuronal differentiation in the hippocampal dentate gyrus in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | KHS 101 hydrochloride |
| Purity | >98% |
| Molecular Weight (g/mol) | 375.92 |
| Inhibitors | KHS 101 hydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction