missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KIAA1018 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | KIAA1018 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC047882 |
| Molecular Weight (g/mol) | Theoretical M.W. 59.6kDa |
| Plasmid | pCMV-KIAA1018 full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-KIAA1018 antibody (Mfr. No. H00022909-B01) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?