missing translation for 'onlineSavingsMsg'
Learn More

Kieselguhr, Pure, Washed with Acid, Fisher Chemical™

Product Code. 10385850 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Keg
Plastic Powder Jar
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10385850 500 g Plastic Powder Jar 500g
10763991 1 kg Plastic Keg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10385850 Supplier Fisher Chemical Supplier No. K/0100/53
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

Physical Form Solid
Color Gray
Quantity 500 g
pH 9 to 10
CAS 61790-53-2
Packaging Plastic Powder Jar
Molecular Formula O2Si
MDL Number 132803
compliance-icons compliance-icons
Product Identifier
  • Kieselguhr
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H373-May cause damage to organs.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.