missing translation for 'onlineSavingsMsg'
Learn More

Kieselguhr, Pure, White, Fisher Chemical

Product Code. 10234200 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10234200 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10234200 Supplier Fisher Chemical Supplier No. K/0090/60
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Physical Form Powder Solid
Color White
Quantity 1 kg
CAS 61790-53-2
Packaging Plastic Powder Jar
Molecular Formula O2Si
MDL Number 132803
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.