missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Kifunensine

Product Code. 18064762
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18064762 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18064762 Supplier R&D Systems™ Supplier No. 3207/1
  • ONLINE EXCLUSIVE PRICE
    231.22 €
    206.85 € / 1mg
    Save 24.37 €
    10% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of class I α-mannosidases

Kifunensine is an inhibitor of class I α-mannosidases that inhibits glycoprotein processing. Kifunensine inhibits human endoplasmic reticulum α-1,2-mannosidase I and Golgi Class I mannosidases IA, IB and IC with Ki values of 130 and 23 nM respectively. Kifunensine disrupts normal trafficking of the GLUT1 transporter, leading to decreased glucose uptake in human renal epithelial cells. Kifunensine impaires tumour cell aggregation in human ovarian cancer cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

CAS 109944-15-2
Quantity 1 mg
Product Line Tocris
Molecular Formula C8H12N2O6
Formula Weight 232.19
Purity >98%
Molecular Weight (g/mol) 232.192
Chemical Name or Material Kifunensine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.