missing translation for 'onlineSavingsMsg'
Learn More

Cytiva KiloBase DNA Marker

Product Code. 11545115 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity unitSize
11545115 50 μg 50µg
1 options
This item is not returnable. View return policy

Product Code. 11545115

Brand: Cytiva 27400401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for quick and precise sizing of double-stranded DNA fragments from 500 bp to 10.0 kb on agarose gels.

  • Bands stain with approximately the same intensity, except for the 1.0-kb band, which is three times the intensity of the other bands
  • Consists of 11 easily-identifiable bands at 0.5, 1.0, 1.5, 2.0, 2.5, 3.0, 4.0, 5.0, 6.0, 8.0, and 10.0 kb when run on a 0.8% agarose ge
  • Not designed for quantitating DNA content in a sample
  • Supplied at 0.5 μg/°l in TE buffer (pH 8.0) in sufficient quantity for 100 loadings using 1 °l per loading after mixing with tracking dye
  • For quick and precise sizing of double-stranded DNA fragments from 500 bp to 10.0 kb on agarose gels

Not intended for use in quantitative analysis

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 μg/μL
Form Liquid
Formulation 0.5μg/μL in TE Buffer (pH 8) in sufficient quantity for 100 loadings using 1μL per loading after mixing with tracking dye
For Use With (Application) Quick and precise sizing of double-stranded DNA fragments from 500bp to 10kb on Agarose gels
No. of Bands 11
Quantity 50 μg
Product Type DNA Molecular Weight Marker/Ladder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.