missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ G60 Purple Nitrile™ Cut-Resistant Gloves

Product Code. 11893480
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11893480 9 12 pairs
11873480 7 12 pairs
11883480 8 12 pairs
11803490 10 12 pairs
11710674 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11893480 Supplier KleenGuard™ Supplier No. 97432
  • ONLINE EXCLUSIVE PRICE
    242.90 €
    136.75 € / 12 pairs
    Save 106.15 €
    43% Off
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cut-resistant gloves are constructed of comfortable, breathable fabric and feature purple tapered coating on the fingertips for added dexterity.

High level of cut resistance while enhancing dexterity, grip and comfort.
  • Dyneema™ fabric construction
  • Purple Nitrile coated fingertips
  • Flexible dotted palm
  • Ambidextrous
TRUSTED_SUSTAINABILITY

Specifications

Coating Material Nitrile Dots
Color White, Purple
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Latex Free Yes
Material Dyneema
Product Type Cut-resistant Gloves
Size 9
EN388 Standard EN 388
For Use With (Application) Glass/Metal handling, Paper making and automotive/aircraft assembly
Glove Cuff Style Knit
Model 97432
Series Kleenguard™ G60
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.