missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Black Phenolic Screw-Thread Closures

Product Code. 10455342
Change view
Click to view available options
Neck Finish:
13-415
15-415
Unit Size:
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish unitSize
10455342 15-415 Pack of 500
11894005 13-415 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10455342 Supplier DWK Life Sciences Supplier No. 7380215415
Trusted Glassware Deals
Use promo code 28509
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

Highly resistant to temperature and steam during autoclaving

  • Cemented-in PTFE-faced 14B white rubber liner
  • Three screw-thread finishes

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
Liner Material PTFE-faced Rubber
Neck Finish 15-415
Closure Included Yes
Product Type Screw Cap
For Use With (Equipment) Bottle
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.