missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ BioMasherII™ Closed System Micro Tissue Homogenizer

Product Code. 13440289
Change view
Click to view available options
Packaging:
Bulk
Single pack
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Sterility Unit Size
13440289 Bulk Non-sterile Pack of 50
15344182 Single pack Sterile Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13440289 Supplier DWK Life Sciences Supplier No. 7496250010
  • ONLINE EXCLUSIVE PRICE
    140.00 €
    119.82 € / Pack of 50
    Save 20.18 €
    14% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensure efficient grinding of samples by minimizing potential exposure to hazardous materials

  • Clear polypropylene tube
  • Polyacetal pestle with molded-in shaft guard
  • Abrasive surface (pestle tip and inner tube area)
  • Available in Sterile and non-sterile versions
  • Capacity: 1.5mL

For animal tissues and organs, plant materials, insects, DNA, RNA, proteins, yeasts and enzymes

Specifications

Capacity (English) 0.05 oz.
For Use With (Application) For animal tissues and organs, plant materials, insects, DNA, RNA, Proteins, yeasts and Enzymes
Autoclavable Autoclavable
Color Clear
Includes PP tube, POM pestle
Capacity (Metric) 1.5 mL
Packaging Bulk
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.