Learn More
DWK Life Sciences KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly Component Parts
Product Code. 11803123
Change view
Click to view available options
Type:
Clamp
Unit Size:
Each
Product Code. Type Unit Size
11803123 Clamp Each
1 options
Product Code. 11803123 Supplier DWK Life Sciences Supplier No. 9537530000
  • ONLINE EXCLUSIVE PRICE
    123.00 €
    113.90 € / Pack of 1
    Save 9.10 €
    7% Off
Estimated Shipment: 17-09-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
KIMBLE™ KONTES™ Ultra-Ware™ Microfiltration Assembly Component Parts are replacement and accessory components designed for use with the Ultra-Ware™ microfiltration assembly system, ensuring reliable filtration performance in laboratory applications.
  • Replacement and accessory components for Kimble™ Kontes™ Ultra-Ware™ microfiltration assemblies
  • Includes glass funnels, support bases, and aluminum clamps to maintain or expand filtration setups
  • Support bases available in 47mm and 90mm sizes to accommodate different filter membrane diameters
  • Aluminum clamp provides secure sealing of funnel to support base during filtration
  • Designed for compatibility with standard Ultra-Ware™ microfiltration assembly configurations
  • Suitable for laboratory filtration of aqueous and compatible solvent samples
Specifications
Disposable Reusable
Material Anodized Aluminum
Sterility Non-sterile
Type Clamp
For Use With (Equipment) Ultra-ware microfiltration assembly with PTFE-faced fritted glass support