Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column ETFE Ferrule for 1/8 in. Tubing
Product Code. 11792304 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
PTFE
Unit Size:
Each
Product Code. Material Unit Size
11792304 PTFE Each
1 options
Product Code. 11792304 Supplier DWK Life Sciences Supplier No. 4208220018
  • ONLINE EXCLUSIVE PRICE
    5.30 €
    4.12 € / Pack of 1
    Save 1.18 €
    22% Off
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™, DISPOSAFLEX™ columns, and ULTRA-WARE™ filtration assemblies.
  • EFTE Flangeless Ferrule for 1/8 in. OD Tubing
  • For use with 420821 Series Male Nuts
  • Case of 10
  • Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
Specifications
Type Ferrule
For Use With (Equipment) HPLC cap system
Material PTFE
Quantity 1 Cs.