missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ Packing Reservoir for CHROMAFLEX™ Columns

Product Code. 15310168
Change view
Click to view available options
Diameter (Metric):
1 cm
2.5 cm
Quantity:
150 mL
500 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Quantity unitSize
15310168 1 cm 150 mL Each
15350168 2.5 cm 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15310168 Supplier DWK Life Sciences Supplier No. 4208370000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHROMAFLEX™ packing reservoirs are ideal for packing gel slurries into columns

  • Fits Standard Column
  • Each packing reservoir is designed for easy mounting on the CHROMAFLEX™ columns
  • The capacities are matched to take the amount of gel slurry required to pack the longest column
  • The packing reservoirs may also be used as buffer reservoirs for gravity flow chromatography
  • 1.0cmID column has nominal volume of 150mL
  • 2.5cmID column has nominal volume of 500mL
  • Temperature Range 0 to 50°C
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY

Specifications

Packaging End fitting has 1/4-28 threads for easy connection to pumps
Quantity 150 mL
Type Packing Reservoir
Diameter (Metric) 1 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.