Learn More
DWK Life Sciences KIMBLE™ Closures for 20mL Glass and Plastic Scintillation Vials
Product Code. 11299819
Change view
Click to view available options
Material (Liner):
Polyethylene
Closure Material:
Urea
Unit Size:
Pack of 1000
Product Code. Material (Liner) Closure Material Unit Size
11299819 Polyethylene Urea Pack of 1000
1 options
Product Code. 11299819 Supplier DWK Life Sciences Supplier No. 7452522400
  • ONLINE EXCLUSIVE PRICE
    261.00 €
    231.94 € / Pack of 1000
    Save 29.06 €
    11% Off
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
KIMBLE™ Closures for 20mL Glass and Plastic Scintillation Vials are durable screw thread closures compatible with 20mL Fisherbrand™ and KIMBLE™ scintillation vials. Available in polyethylene and urea materials with GPI 22-400 thread for a secure, reliable seal.
  • Designed for use with 20mL Fisherbrand™ and Kimble scintillation vials (except 03-340-4B and 03-340-129)
  • GPI 22-400 screw thread finish for secure closure
  • Available in white polyethylene (linerless) and white urea constructions
  • Liner options include no liner, cone-shaped polyethylene, and PTFE-faced white rubber
  • Top surface suitable for marking
  • Not autoclavable
Specifications
Material (Liner) Polyethylene
Color White
Material Urea
Closure Material Urea
Closure Type Screw
Quantity 1000/Cs.
Autoclavable Not Autoclavable
Product Type Screw Cap