Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
Click to view available options
Closure Size:
15-425
18-400
20-400
24-400
Unit Size:
Pack of 144
Description
Black phenolic closure with PTFE-lined cap
- Works with media bottles and screw-thread vials
- Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
- Cemented-in PTFE-faced rubber liner is provided
Specifications
Specifications
| Closure Size | 15-425 |
| Liner Material | PTFE, Rubber |
| Product Type | Screw Cap |
| Material | Phenolic |
| Color | Black |