Learn More
DWK Life Sciences™ Kimble™ Glass Racks for Westergren Tubes
Product Code. 15929665
Change view
Click to view available options
Positions:
10
For Use With (Application):
Westergren and Wintrobe Tubes
Unit Size:
Each
Product Code. Positions For Use With (Application) unitSize
15929665 10 Westergren and Wintrobe Tubes Each
1 options
Product Code. 15929665 Supplier DWK Life Sciences™ Supplier No. 2215N
Only null left
Add to basket
Add to basket
Reduces the chance of bio hazardous contamination
  • Racks feature leveling bubbles and adjustment screws
  • Tubes are held firmly in a perpendicular position to ensure accurate readings.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Material Glass
Positions 10
For Use With (Application) Westergren and Wintrobe Tubes