missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Kontes™ TLC Cylindrical Developing Tanks

Product Code. 12746728 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Material:
Glass
Holds:
5 L x 10 cm W Plate
Quantity:
1/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Material Holds Quantity Unit Size
12746728 Glass 5 L x 10 cm W Plate 1/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12746728 Supplier DWK Life Sciences Supplier No. 4161701120
  • ONLINE EXCLUSIVE PRICE
    104.00 €
    84.62 € / Pack of 1
    Save 19.38 €
    18% Off
 Restricted Item
Estimated Shipment: 20-05-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cylindrical glass tanks for the development of 5 x 10cm (2 x 3.9 in. ) and 5 x 20cm (2 x 7.9 in. ) plates

Tanks provide maximum vapor phase saturation with a minimum amount of solvent

Specifications

Closure Material Glass
Material Borosilicate Glass
Quantity 1/Pk.
Type Cylindrical TLC Developing Tank
For Use With (Application) Provides Maximum Vapor Phase Saturation with a Minimum Amount of Solvent
Holds 5 L x 10 cm W Plate
Shape Cylindrical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.