missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ Linear HDPE Stoppers
Click to view available options
Size:
13
Stopper Number:
13
Unit Size:
Each
Description
Closed bottom end fits the standard taper neck
- Heavy-duty Polyethylene stopper
- Flange on stopper protects glass
Specifications
Specifications
| For Use With (Application) | Used as replacement parts for items that are ground to standard taper stopper dimensions |
| Size | 13 |
| Color | Yellow, Blue |
| Material | HDPE |
| Product Type | Tapered Stopper |
| Stopper Number | 13 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction