missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ Serum KG-35 Borosilicate Glass Bottle, Aluminum Seal, No Closure

Product Code. 15219383
Change view
Click to view available options
Capacity (Metric):
200 mL
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15219383 200 mL Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15219383 Supplier DWK Life Sciences Supplier No. 61000G 200

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KIMBLE™ Serum KG-35 Borosilicate Glass Bottle, Aluminum Seal, No Closure is made from USP Type I borosilicate glass. Ideal for injectable and parenteral solutions. Highly resistant to thermal, mechanical shock, and chemical attack.

Neutraglas™

  • Manufactured from USP Type I borosilicate molded glass
  • Ideal for packaging and storage of injectable and parenteral solutions requiring utmost purity
  • Highly resistant to thermal and mechanical shock as well as chemical attack
  • GPI Neck Finish: 20A
  • Supplied without closure

Specifications

Capacity 6.76 oz., 200mL
Capacity (Metric) 200 mL
Color Clear
Diameter (English) 2.55 in.
Material Borosilicate Glass
Length (Metric) 114mm
Sterility Non-sterile
Mouth Narrow
Length (English) 4.48 in.
Closure Material Aluminum
Closure Size 20A
Diameter (Metric) 65 mm
Capacity (English) 6.76 oz.
Shape Round
Product Type Serum Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.