missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ Straight-Sided Serum Stoppers

Product Code. 10632174 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Outer:
20 mm
Quantity:
100/Cs.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Quantity Unit Size
10632174 20 mm 100/Cs. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10632174 Supplier DWK Life Sciences Supplier No. 73811T21
  • ONLINE EXCLUSIVE PRICE
    123.00 €
    109.06 € / Pack of 100
    Save 13.94 €
    11% Off
 Restricted Item
Estimated Shipment: 18-06-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

KIMBLE™ Straight-Sided Serum Stoppers are designed for research and pharmaceutical packaging applications. The universal gray chlorobutyl formulation meets USP, EP, and JP requirements and is free of plasticizers, nitrosamine precursors, and natural rubber latex.

  • For use with 13mm and 20mm serum-finished bottles and vials
  • Universal gray chlorobutyl formulation meets USP, EP, and JP requirements for globally marketed pharmaceutical products
  • Contain no plasticizers, 2-mercapto-benzothiazole, nitrosamine precursors, or natural rubber latex
  • Applicable to aqueous solutions with a pH range of 2 to 10
  • PTFE facing (73811T series) improves chemical compatibility and minimizes surface interactions
  • Durometer 50

Order Info

Durometer 50.

Specifications

Certifications/Compliance USP, EP, JP
Description Latex free
For Use With (Application) For research and pharmaceutical packaging applications
For Use With (Equipment) 20 mm GPI Aluminum Seal
Hardness 50 Durometer
Quantity 100/Cs.
Autoclavable Autoclavable
Color Gray
Diameter (Metric) Bottom 13 mm
Diameter (Metric) Outer 20 mm
Diameter (Metric) Top 20 mm
Material Chlorobutyl Rubber, PTFE
Product Type Straight Plug Stopper
Shape Cylindrical
Sterility Non-sterile
Stopper Number 20
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.