Learn More
Kimtech™ G3 Latex Ambidextrous Gloves
Product Code. 11690423
Change view
Click to view available options
Size:
Large
Unit Size:
Pack of 1000
Product Code. Size unitSize
11690423 Large Pack of 1000
1 options
Product Code. 11690423 Supplier Kimtech™ Supplier No. HC445
Only null left
Add to basket
Add to basket
G3 latex cleanroom gloves provide sensitive and dextrous protection in ISO Class 3 or higher cleanrooms. Textured finish for enhanced tactile performance.
  • Assured Compliance:
    • PPE Cat III according to Regulation (EU) 2016/425
    • EN ISO 374-1:2016 Type C (K) Chemical Splash protection
    • EN 374-4:2003 Resistance to degradation by chemicals
    • EN ISO 374-5:2016 Micro Organism and Virus Protection
    • Certificate of Analysis available online
  • High quality latex material provides high levels of contamination protection against micro-organisms, chemical splash, particles and extractables.
  • Suitable for use in ISO Class 3 or higher cleanrooms, and provided with supporting technical information to demonstrate regulatory compliance.
  • Gloves are non-sterile, ambidextrous, disposable and a natural color.
  • Textured palm and fingertips enhance grip and tactile sensitivity for safer and more efficient processes.
  • Beaded cuffs add strength to the gloves, reducing the risk of tearing and increasing their durability, while also reducing roll down for easier donning and doffing.
  • Made from natural rubber latex washed repeatedly in ultrapure deionized water and containing no powder, reducing the risks of skin irritation for the wearer.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001; CE CAT III; Category III PPE - CE Complex
Color Natural
Exterior Finish Fully Textured
Hand Ambidextrous
Material Latex
Powdered Powder Free
Size Large
Sterility Non-sterile
Glove Cuff Style Beaded
Length (Metric) 30 cm