missing translation for 'onlineSavingsMsg'
Learn More

Kimtech™ Opal™ Nitrile Gloves
GREENER_CHOICE

Product Code. 17307669
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 170
Pack of 200
This item is not returnable. View return policy

Product Code. 17307669

Brand: Kimtech™ 62884

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

These powder free, ambidextrous Kimtech™ Opal™ Nitrile Gloves with beaded cuff and textured fingertips guard against contamination by chemical splash and micro-organism hazards while reducing the risk of TYPE I and IV glove-associated skin reactions.

  • Kimtech™ Opal™ nitrile gloves guard against contamination by chemical splash and micro-organism hazards, delivering seamless protection when and where it counts.
  • The powder-free gloves are ideal for use in research and production facilities, life sciences and non-sterile drug manufacturing applications.
  • The gloves feature an innovative approach using a thin nitrile polymer with a tip thickness of just 0.07 mm, but with excellent tensile strength suitable for rigorous process use.
  • Easy to don, soft and comfortable Kimtech™ Opal™ nitrile gloves are suitable for extended wearing time.
  • The absence of natural rubber latex, sulphur and vulcanizing agents* reduces the risk of TYPE I and TYPE IV glove-associated skin reactions, protecting the wearer as well as the application.
  • Precision manufacturing reduces the risk of contamination and ensures the nitrile gloves offer high levels of regulatory compliance.
  • Beaded cuffs and textured fingertips enable excellent handling of both wet and dry materials, and the ambidextrous gloves have been extensively tested under stringent conditions.
  • In addition, the thin nitrile construction and efficient packaging mean that the chemical-resistant gloves can reduce waste by up to 50%.
  • Available in multiple sizes: X-Small (62880), Small (62881), Medium (62883), Large (62883), X-Large (62884)

*No sulphur added and formulated without the following vulcanising chemicals and accelerators: Thiurams, Thiazoles, Guanidines and Carbamates.

Specifications

Certifications/Compliance ISO 3
Certifications/Compliance (Europe) PPE Cat III according to Regulation (EU) 2016/425, EN ISO 374-1 Type B, EN ISO 374-5 Micro Organism and Virus Protection
Accelerator Free Yes
AQL 1.5
Color Dark Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 1.8 mil
Finger Thickness (Metric) 0.18 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2 mil
Palm Thickness (Metric) 0.05 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Large
Sterility Non-sterile
Thickness (English) 2 mil
Thickness (Metric) 0.05 mm
Length (English) 9.45 in.
Length (Metric) 24 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.